6-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Amino-4-methyl-2H-benzo[b][1,4]oxazin-3(4H)-one

A165550-100MG A165550-1G A165550-250MG A165550-5G
ALA165550100MGEA 15.11 USD
ALA165550100MG ALA165550-1G ALA165550250MG ALA165550-5G
6-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥97%
6-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one
Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01834021
CAS Number: 103361-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2083ae5667b50f50f850eed60bc86771ad075394&vid=2083ae5667b50f50f850eed60bc86771ad075394&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR