Adenosine 5′-diphosphate bis(cyclohexylammonium) salt ≥95% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: ADP

A165484-1G A165484-250MG
ALA165484-1GEA 499.61 USD
ALA165484-1G ALA165484250MG
Adenosine 5′-diphosphate bis(cyclohexylammonium) salt ≥95% (by HPLC)
Adenosine 5′-diphosphate bis(cyclohexylammonium) salt

Adenosine 5 '- diphosphate (ADP) is an adenine nucleotide involved in energy storage and nucleic acid metabolism through the conversion of ATP synthase to ATP. ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12, and P2X1. After being converted to adenosine through the ecto ADP enzyme, platelet activation is inhibited through adenosine receptors.

Formula: C₁₀H₁₅N₅O₁₀P₂ · 2C₆H₁₃N
MW: 625.55 g/mol
Storage Temperature: Freezer
CAS Number: 102029-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e556fb7f5f7ec632207592aecb348789345dc26b&vid=e556fb7f5f7ec632207592aecb348789345dc26b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR