2-Amino-3-methoxypyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A165482-1G A165482-5G
ALA165482-1GEA 13.21 USD
ALA165482-1G ALA165482-5G
2-Amino-3-methoxypyridine ≥97%
2-Amino-3-methoxypyridine
Formula: C₆H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD07374874
CAS Number: 10201-71-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cec696a8b42125e3f79b24282c539080d84cef4&vid=4cec696a8b42125e3f79b24282c539080d84cef4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR