5-Amino-2-ethoxybenzonitrile 97

Supplier: ALADDIN SCIENTIFIC

A165471-1G A165471-250MG
ALA165471-1GEA 201.31 USD
ALA165471-1G ALA165471250MG
5-Amino-2-ethoxybenzonitrile 97
5-Amino-2-ethoxybenzonitrile
Formula: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD10686562
CAS Number: 1020046-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67af4dc81ba346324d75c83051a3bc1132a7696d&vid=67af4dc81ba346324d75c83051a3bc1132a7696d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR