3-Amino-5-bromopyridin-4-ol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-5-bromo-4-hydroxypyridine

A165392-1G A165392-250MG A165392-50MG A165392-5G
ALA165392-1GEA 575.61 USD
ALA165392-1G ALA165392250MG ALA165392-50MG ALA165392-5G
3-Amino-5-bromopyridin-4-ol ≥97%
3-Amino-5-bromopyridin-4-ol
Formula: C₅H₅BrN₂O
MW: 189.012 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08692340
CAS Number: 101084-20-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14b4c8a9abd04ca9a552bbf157e337bdf8b85129&vid=14b4c8a9abd04ca9a552bbf157e337bdf8b85129&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR