2-Aminothiazole-5-carbaldehyde ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Aminothiazole-5-carboxaldehyde, 2-Amino-1,3-thiazole-5-carboxaldehyde

A165357-1G A165357-5G
ALA165357-1GEA 110.11 USD
ALA165357-1G ALA165357-5G
2-Aminothiazole-5-carbaldehyde ≥95%
2-Aminothiazole-5-carbaldehyde

The presence of different functional groups allows differential attachment of an azo linkage and benzoxazolium salt providing cationic, delocalized azo dyes.

Formula: C₄H₄N₂OS
MW: 128.15 g/mol
Melting Pt: 160°C
Storage Temperature: Ambient
MDL Number: MFCD02179570
CAS Number: 1003-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be8669a46a4b5452f8f6b93e6c529f4b5370934c&vid=be8669a46a4b5452f8f6b93e6c529f4b5370934c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR