3-Amino-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 3-Amino-4-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Amino-4-fluorophenylboronic acid, pinacol ester

A165356-1G A165356-250MG A165356-100MG
ALA165356-1GEA 126.26 USD
ALA165356-1G ALA165356250MG ALA165356100MG
3-Amino-4-fluorophenylboronic acid pinacol ester ≥97%
3-Amino-4-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD11044430
CAS Number: 1003575-43-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=031116c8a8693cb7fe5d334b58af2e2daa233396&vid=031116c8a8693cb7fe5d334b58af2e2daa233396&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR