8-Aminocaprylic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: ω-Aminooctoic acid, 8-Aminooctanoic acid, ω-Aminocaprylic acid, ω-Aminooctanoic acid, 8-Aminooctoic acid

A165345-100G A165345-1G A165345-25G A165345-5G
ALA165345-100GEA 2316.01 USD
ALA165345-100G ALA165345-1G ALA165345-25G ALA165345-5G
8-Aminocaprylic acid ≥98%
8-Aminocaprylic acid

Application:8-Aminooctanoic Acid is a substance used in the study on the activity and specificity of ω-aminooctanoic acid-glycine-aspartic acid-tryptophan peptide in inhibiting platelet aggregation. Study on lysine analog effect on plasminogen binding in PC12 cells.

Formula: C₈H₁₇NO₂
MW: 159.23 g/mol
Melting Pt: 194°C
Storage Temperature: Ambient
MDL Number: MFCD00008245
CAS Number: 1002-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70336558d05bce6d71e0ca78de107e045dbf5765&vid=70336558d05bce6d71e0ca78de107e045dbf5765&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR