2-(Aminomethyl)benzoic acid hydrochloride ≥95%

Supplier: Aladdin Scientific
Danger

A165340-1G A165340-250MG A165340-5G
ALA165340-1GEA 354.26 USD
ALA165340-1G ALA165340250MG ALA165340-5G
2-(Aminomethyl)benzoic acid hydrochloride ≥95%
2-(Aminomethyl)benzoic acid hydrochloride
Formula: C₈H₁₀ClNO₂
Melting Pt: 219-225 °C
Storage Temperature: Ambient
MDL Number: MFCD09028989
CAS Number: 10017-39-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=904e22179c133378c22d178d764fda72d54897ce&vid=904e22179c133378c22d178d764fda72d54897ce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR