Pentyl nitrate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Pentyl nitrate, Amyl nitrate

A165297-1ML A165297-5ML
ALA165297-1MLEA 68.31 USD
ALA165297-1ML ALA165297-5ML
Pentyl nitrate ≥98% (by GC)
Pentyl nitrate
Formula: C₅H₁₁NO₃
MW: 133.15 g/mol
Boiling Pt: 158
Melting Pt: -123
Flash Pt: 52°C
Storage Temperature: Ambient
MDL Number: MFCD00059177
CAS Number: 1002-16-0
UN: 1112
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d0aa39c979da12368f1355f1d6e896cffc9fb4e&vid=1d0aa39c979da12368f1355f1d6e896cffc9fb4e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR