Azido PEG
Supplier: Aladdin Scientific
A163315-1G A163315-250MG
ALA163315-1GEA 849.21 USD
ALA163315-1G ALA163315250MG
Azido PEG
Azido PEG
Azide/azido functionalized methoxyl polyethylene glycol (mPEG-N3) is a monofunctional PEG derivative that can be used to modify proteins, peptides and other materials. Azide group can react with alkyne in aqueous solution catalyzed by copper. It can also be reduced into amine group easily. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.