Azido PEG NHS
Supplier: ALADDIN SCIENTIFIC
Synonyms: N3-PEG-NHS
A163304-100MG A163304-500MG
ALA163304100MGEA 164.26 USD
ALA163304100MG ALA163304500MG
Azido PEG NHS
Azido PEG NHS
Azide/azido functionalized polyethylene glycol (N3-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials. Azide group can react with alkyne in aqueous solution catalyzed by copper. It can also be reduced into amine group easily while another functional group can be react with other molecules or functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.