Azido PEG NHS
Supplier: Aladdin Scientific
A163263-1G A163263-250MG
ALA163263-1GEA 164.26 USD
ALA163263-1G ALA163263250MG
Azido PEG NHS
Azido PEG NHS
Amino functionalized methoxyl polyethylene glycol, amino PEG (mPEG-NH2) is a mono reactive PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.