Azido PEG NHS
Supplier: Aladdin Scientific
A163260-100MG A163260-1G A163260-500MG
ALA163260100MGEA 86.36 USD
ALA163260100MG ALA163260-1G ALA163260500MG
Azido PEG NHS
Azido PEG NHS
Amino functionalized methoxyl polyethylene glycol, amino PEG (mPEG-NH2) is a mono reactive PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.