Azido PEG NHS ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: N3-PEG-NHS
A163259-100MG A163259-1G A163259-5G
ALA163259100MGEA 98.71 USD
ALA163259100MG ALA163259-1G ALA163259-5G
Azido PEG NHS ≥95%
Azido PEG NHS
Amino functionalized methoxyl polyethylene glycol, amino PEG (mPEG-NH2) is a mono reactive PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.