Azido PEG amine

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N3-PEG-NH2

A163257-1G A163257-200MG A163257-50MG
ALA163257-1GEA 1045.86 USD
ALA163257-1G ALA163257200MG ALA163257-50MG
Azido PEG amine
Azido PEG amine

Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.

Formula: NH₂(CH₂CH₂O)nH
Storage Temperature: Ambient
CAS Number: 32130-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR