Aspartic Acid PEG Aspartic Acid
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163249-1G
A163249-250MG
ALA163249-1GEA
229.81
USD
ALA163249-1G
ALA163249250MG
Aspartic Acid PEG Aspartic Acid
Aspartic Acid PEG Aspartic Acid
Amino functionalized polyethylene glycol (NH2-PEG-NH2) is a homobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.