Amino PEG
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
mPEG-NH2
A163247-1G
A163247-200MG
ALA163247-1GEA
229.81
USD
ALA163247-1G
ALA163247200MG
Amino PEG
Amino PEG
Amino functionalized polyethylene glycol (NH2-PEG-NH2) is a homobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.