Amino PEG hydroxyl
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: NH2-PEG-OH
A163243-1G A163243-250MG
ALA163243-1GEA 187.06 USD
ALA163243-1G ALA163243250MG
Amino PEG hydroxyl
Amino PEG hydroxyl
Amino functionalized polyethylene glycol (NH2-PEG-NH2) is a homobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.