Amino PEG hydroxyl
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: NH2-PEG-OH
A163242-250MG A163242-5G
ALA163242250MGEA 22.71 USD
ALA163242250MG ALA163242-5G
Amino PEG hydroxyl
Amino PEG hydroxyl
Amino functionalized polyethylene glycol (NH2-PEG-NH2) is a homobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.