Amino PEG Amine
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: NH2-PEG-NH2
A163235-100MG A163235-500MG
ALA163235100MGEA 229.81 USD
ALA163235100MG ALA163235500MG
Amino PEG Amine
Amino PEG Amine
Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.