Amino PEG Acid
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: NH2-PEG-COOH
A163227-100MG A163227-500MG
ALA163227100MGEA 196.56 USD
ALA163227100MG ALA163227500MG
Amino PEG Acid
Amino PEG Acid
Alkyne functionalized polyethylene glycol (Alk-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials via Click Chemistry. Alkyne group can react with azide in aqueous solution catalyzed by copper. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.