Amino PEG Acid
Supplier: ALADDIN SCIENTIFIC
Synonyms: NH2-PEG-COOH
A163225-100MG A163225-500MG
ALA163225100MGEA 164.26 USD
ALA163225100MG ALA163225500MG
Amino PEG Acid
Amino PEG Acid
Alkyne functionalized polyethylene glycol (Alk-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials via Click Chemistry. Alkyne group can react with azide in aqueous solution catalyzed by copper. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.