Amino PEG Acid
Supplier: ALADDIN SCIENTIFIC
A163222-100MG A163222-500MG
ALA163222100MGEA 170.91 USD
ALA163222100MG ALA163222500MG
Amino PEG Acid
Amino PEG Acid
Alkyne functionalized polyethylene glycol (Alk-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials via Click Chemistry. Alkyne group can react with azide in aqueous solution catalyzed by copper. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.