Alkyne PEG NHS
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: ALK-PEG-NHS
A163212-1G A163212-500MG A163212-100MG
ALA163212-1GEA 751.36 USD
ALA163212-1G ALA163212500MG ALA163212100MG
Alkyne PEG NHS
Alkyne PEG NHS
Alkyne functionalized polyethylene glycol (Alk-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials via Click Chemistry. Alkyne group can react with azide in aqueous solution catalyzed by copper. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.