Aldehyde PEG Aldehyde
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
CHO-PEG-CHO
A163180-100MG
A163180-500MG
ALA163180100MGEA
326.71
USD
ALA163180100MG
ALA163180500MG
Aldehyde PEG Aldehyde
Aldehyde PEG Aldehyde
Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.