Acryloyl-PEG-NHS
Supplier: ALADDIN SCIENTIFIC
A163178-100MG A163178-500MG
ALA163178100MGEA 248.81 USD
ALA163178100MG ALA163178500MG
Acryloyl-PEG-NHS
Acryloyl-PEG-NHS
Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.