Acryloyl PEG amine
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163166-100MG A163166-500MG
ALA163166100MGEA 262.11 USD
ALA163166100MG ALA163166500MG
Acryloyl PEG amine
Acryloyl PEG amine
Aacrylate functionalized PEG derivatives can be used to make hydrogel via radical polymerization process. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .