Acrylate PEG Biotin ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163146-500MG
A163146-100MG
ALA163146500MGEA
292.9
USD
ALA163146500MG
ALA163146100MG
Acrylate PEG Biotin ≥95%
Acrylate PEG Biotin
8 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .