8-Arm PEG Amine
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163131-100MG A163131-25MG
ALA163131100MGEA 138.9 USD
ALA163131100MG ALA163131-25MG
8-Arm PEG Amine
8-Arm PEG Amine
8 arm PEG biotin derivatives have 8 biotins one each PEG molecule that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .