4-Arm PEG vinyl sulphone
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163123-100MG
A163123-500MG
ALA163123100MGEA
240.9
USD
ALA163123100MG
ALA163123500MG
4-Arm PEG vinyl sulphone
4-Arm PEG vinyl sulphone
4 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .