4-Arm PEG Amine
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms: 4 Arm PEG Amine
A163115-1G A163115-250MG
ALA163115-1GEA 327.66 USD
ALA163115-1G ALA163115250MG
4-Arm PEG Amine
4-Arm PEG Amine
4 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .