4-Arm PEG DTPA
Supplier: Aladdin Scientific
A163111-100MG A163111-500MG
ALA163111100MGEA 128.16 USD
ALA163111100MG ALA163111500MG
4-Arm PEG DTPA
4-Arm PEG DTPA
4 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .