4-Arm PEG NHS ≥95%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
4arm-PEG10K-Succinimidyl Carboxymethyl Ester, 4 Arm PEG NHS, 4arm-PEG-Succinimidyl Carboxymethyl Ester, 4arm-PEG-SCM
A163109-100MG
A163109-500MG
ALA163109100MGEA
164.26
USD
ALA163109100MG
ALA163109500MG
4-Arm PEG NHS ≥95%
4-Arm PEG NHS
4arm PEG SCM (4arm PEG NHS Ester). Hydrogel PEG. This is the activated form of 4ARM-COOH.Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization