2-Arm/Branched PEG NHS
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163093-100MG A163093-1G A163093-500MG
ALA163093100MGEA 110.9 USD
ALA163093100MG ALA163093-1G ALA163093500MG
2-Arm/Branched PEG NHS
2-Arm/Branched PEG NHS
2 arm PEG derivatives have two PEG branches with one or two functional groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .