2-Arm/Branched PEG Biotin
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163089-100MG A163089-500MG
ALA163089100MGEA 137.9 USD
ALA163089100MG ALA163089500MG
2-Arm/Branched PEG Biotin
2-Arm/Branched PEG Biotin
2 arm PEG derivatives have two PEG branches with one or two functional groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .