2-Arm/Branched PEG Amine
Supplier: ALADDIN SCIENTIFIC CORPORATION
A163088-100MG A163088-500MG
ALA163088100MGEA 137.9 USD
ALA163088100MG ALA163088500MG
2-Arm/Branched PEG Amine
(1S)-1-Cyclobutylethan-1-amine hydrochloride
2 arm PEG derivatives have two PEG branches with one or two functional groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .