2-Acetamido-6-aminopyridine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A151791-1G A151791-25G A151791-5G
ALA151791-1GEA 20.81 USD
ALA151791-1G ALA151791-25G ALA151791-5G
2-Acetamido-6-aminopyridine ≥98%
2-Acetamido-6-aminopyridine
Formula: C₇H₉N₃O
Melting Pt: 160°C
MDL Number: MFCD01099052
CAS Number: 1075-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2621a393479ea474bd978bb10cbb46c3b9f80e&vid=ee2621a393479ea474bd978bb10cbb46c3b9f80e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR