2-Acetamidopyridine 98 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Warning

A151788-100G A151788-1G A151788-25G A151788-5G
ALA151788-100GEA 3479.76 USD
ALA151788-100G ALA151788-1G ALA151788-25G ALA151788-5G
2-Acetamidopyridine 98 GC_ASSAY_METHOD
2-Acetamidopyridine
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 148 °C
Melting Pt: 69 °C
MDL Number: MFCD00023423
CAS Number: 5231-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9aaa5d98726bc7c8785fb62464b26d9e8438c16a&vid=9aaa5d98726bc7c8785fb62464b26d9e8438c16a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR