3-(2-Aminoethyl)pyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-(Pyridin-3-yl)ethanamine, 2-(3-Pyridyl)ethylamine

A151786-10G A151786-1G A151786-5G
ALA151786-10GEA 159.51 USD
ALA151786-10G ALA151786-1G ALA151786-5G
3-(2-Aminoethyl)pyridine ≥98% (by GC)
3-(2-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 128°C
Density: 1.039 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00191604
CAS Number: 20173-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adc8ddc678e72ad34c45744e2a5a69bed0ccc4c2&vid=adc8ddc678e72ad34c45744e2a5a69bed0ccc4c2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR