3-Pyridyl acetate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151785-25G A151785-5G
ALA151785-25GEA 103.46 USD
ALA151785-25G ALA151785-5G
3-Pyridyl acetate ≥98% (by GC)
3-Pyridyl acetate
Formula: C₇H₇NO₂
Boiling Pt: 137°C
Density: 1.153 g/cm³
Flash Pt: 100°C
MDL Number: MFCD00006377
CAS Number: 17747-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57c8e68fd7dd72093f77cd6582b022601896c725&vid=57c8e68fd7dd72093f77cd6582b022601896c725&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR