1,4-Anhydro-D-xylitol ≥95%

Supplier: ALADDIN SCIENTIFIC

A151783-10MG A151783-25MG A151783-100MG
ALA151783-10MGEA 253.56 USD
ALA151783-10MG ALA151783-25MG ALA151783100MG
1,4-Anhydro-D-xylitol ≥95%
1,4-Anhydro-D-xylitol
Formula: C₅H₁₀O₄
Boiling Pt: 160°C
Flash Pt: 158 °C
Storage Temperature: Refrigerator
CAS Number: 53448-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87b9d7356a64e56d0d980750994d95fe908d25d1&vid=87b9d7356a64e56d0d980750994d95fe908d25d1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR