2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Acetyl-5,5-dimethylcyclohexane-1,3-dione, 2-Acetyldimedone

A151781-1G A151781-250MG A151781-5G
ALA151781-1GEA 33.16 USD
ALA151781-1G ALA151781250MG ALA151781-5G
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione ≥98%
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Melting Pt: 36-40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00467663
CAS Number: 1755-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f990730f2b8775d24557ed79e8a948cafd72cf85&vid=f990730f2b8775d24557ed79e8a948cafd72cf85&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR