1,4-Bis(2-hydroxyisopropyl)benzene ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1,4-Bis(α-hydroxyisopropyl)benzene, 1,4-Bis(1-methyl-1-hydroxyethyl)benzene, 1,4-Bis(alpha-hydroxyisopropyl)benzene, alpha,alpha'-Dihydroxy-1,4-diisopropylbenzene, 2,2'-(p-Phenylene)di-2-propanol, 2,2'-(1,4-Phenylene)bis(propan-2-ol), ^a,^a,^a',^a'-Tetramethyl-1,4-benzenedimethanol, alpha,alpha,alpha',alpha'-Tetramethyl-1,4-benzenedimethanol, α,α,α',α'-Tetramethyl-1,4-benzenedimethanol, α,α'-Dihydroxy-1,4-diisopropylbenzene, 1,4-Bis(2-hydroxyisopropyl)benzene

A151780-100G A151780-1G A151780-25G A151780-5G
ALA151780-100GEA 942.31 USD
ALA151780-100G ALA151780-1G ALA151780-25G ALA151780-5G
1,4-Bis(2-hydroxyisopropyl)benzene ≥97%
1,4-Bis(2-hydroxyisopropyl)benzene
Formula: C₁₂H₁₈O₂
MW: 194.28 g/mol
Boiling Pt: 167-168 °C
Melting Pt: 143-146°C
MDL Number: MFCD00009827
CAS Number: 2948-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb8be9ea4055bc883b00afec1c0b38499d62ea30&vid=eb8be9ea4055bc883b00afec1c0b38499d62ea30&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR