4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid ≥98%

Supplier: Aladdin Scientific
Warning

A151775-100G A151775-1G A151775-25G A151775-5G
ALA151775-100GEA 2310.31 USD
ALA151775-100G ALA151775-1G ALA151775-25G ALA151775-5G
4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid ≥98%
4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid
Formula: C₁₀H₁₃NO₅S
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD04973619
CAS Number: 71675-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d49ba0c0c744e38d5ee9043a8ee03bb21cd5d3ce&vid=d49ba0c0c744e38d5ee9043a8ee03bb21cd5d3ce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR