2'-Amino-4',5'-methylenedioxyacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(6-Aminobenzo[d][1,3]dioxol-5-yl)ethanone, 5-Acetyl-6-amino-1,3-benzodioxole, 6'-Amino-3',4'-(methylenedioxy)acetophenone

A151774-1G A151774-250MG
ALA151774-1GEA 25.56 USD
ALA151774-1G ALA151774250MG
2'-Amino-4',5'-methylenedioxyacetophenone ≥98%
2'-Amino-4',5'-methylenedioxyacetophenone
Formula: C₉H₉NO₃
MW: 179.1727 g/mol
Melting Pt: 170-174°C
MDL Number: MFCD00075855
CAS Number: 28657-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7fb0f971b7ef2831ba6a6915a81ec0b82f066ca&vid=d7fb0f971b7ef2831ba6a6915a81ec0b82f066ca&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR