3-Anilinopropionitrile 98 GC, T

Supplier: ALADDIN SCIENTIFIC
Warning

A151772-100G A151772-25G A151772-5G
ALA151772-100GEA 136.71 USD
ALA151772-100G ALA151772-25G ALA151772-5G
3-Anilinopropionitrile 98 GC, T
3-Anilinopropionitrile
Formula: C₉H₁₀N₂
Boiling Pt: 160°C
Melting Pt: 50 °C
MDL Number: MFCD00001953
CAS Number: 1075-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a2504609809205f7381b067916d67fedfe33607&vid=1a2504609809205f7381b067916d67fedfe33607&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR