4-Aminocinnamic acid ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning
|
Synonyms:
3-(4-Aminophenyl)acrylic acid, 4-Aminocinnamic Acid, 3-(4-Aminophenyl)acrylic Acid
A151768-1G
A151768-25G
A151768-5G
ALA151768-1GEA
29.36
USD
ALA151768-1G
ALA151768-25G
ALA151768-5G
4-Aminocinnamic acid ≥97%
4-Aminocinnamic acid
Formula:
C₉H₉NO₂ MW: 163.18 g/mol |
MDL Number:
MFCD00017117 CAS Number: 2393-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...