(-)-Ambroxide ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning
|
Synonyms:
Ambroxide, (3aR,5aS,9aS,9bR)-3a,6,6,9a-Tetramethyldodecahydronaphtho[2,1-b]furan, 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane
A151758-100G
A151758-1G
A151758-25G
A151758-5G
ALA151758-100GEA
578.46
USD
ALA151758-100G
ALA151758-1G
ALA151758-25G
ALA151758-5G
(-)-Ambroxide ≥98% (by GC)
(-)-Ambroxide
Product Application:(-)-Ambroxide is one of the most important components of natural Ambergris and is useful for making amber fragrance.
Formula:
C₁₆H₂₈O MW: 236.40 g/mol Melting Pt: 77 °C |
MDL Number:
MFCD00134491 CAS Number: 6790-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...