Pentyl hexanoate ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Caproic acid amyl ester, Amyl caproate, Hexanoic acid amyl ester, Caproic acid pentyl ester, Hexanoic acid pentyl ester, Amyl hexanoate, Pentyl hexanoate, Pentyl caproate
A151757-100ML
A151757-25ML
ALA151757100MLEA
104.41
USD
ALA151757100ML
ALA151757-25ML
Pentyl hexanoate ≥98% (by GC)
Pentyl hexanoate
Formula:
C₁₁H₂₂O₂ Boiling Pt: 226 °C Density: 0.867 g/cm³ Flash Pt: 91°C |
MDL Number:
MFCD00027281 CAS Number: 540-07-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...